Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116793
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116793 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116793 Supplier Invitrogen™ Supplier No. PA5116793
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: BxPC-3, Rat lung Immunogen sequence: QVFGDEKKYW LLPIFSSLGD GCSFPTCLVN QDPEQASTPA GLNSTAKNLE NHQFPAKPLR ESQSHLLTDS QSWTESSINP GKCKAGMSNP ALTMENET.

Palmitoyltransferase that catalyzes the addition of palmitate onto various protein substrates and is involved in a variety of cellular processes (PubMed:18508921, PubMed:18296695, PubMed:19144824, PubMed:21343290, PubMed:22034844, PubMed:23793055). In the nervous system, plays a role in long term synaptic potentiation by palmitoylating AKAP5 through which it regulates protein trafficking from the dendritic recycling endosomes to the plasma membrane and controls both structural and functional plasticity at excitatory synapses. In dendrites, mediates the palmitoylation of DLG4 when synaptic activity decreases and induces synaptic clustering of DLG4 and associated AMPA-type glutamate receptors. Also mediates the de novo and turnover palmitoylation of RGS7BP, a shuttle for Gi/o-specific GTPase-activating proteins/GAPs, promoting its localization to the plasma membrane in response to the activation of G protein-coupled receptors. Through the localization of these GTPase-activating proteins/GAPs, it also probably plays a role in G protein-coupled receptors signaling in neurons. Also probably plays a role in cell adhesion by palmitoylating CD9 and CD151 to regulate their expression and function (PubMed:18508921). Palmitoylates the endoplasmic reticulum protein CKAP4 and regulates its localization to the plasma membrane (PubMed:18296695, PubMed:19144824). Could also palmitoylate LCK and regulate its localization to the plasma membrane (PubMed:22034844). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZDHHC2
Gene Accession No. Q9JKR5, Q9UIJ5
Gene Alias 5730415P04Rik; 6430583A19Rik; DHHC2; DHHC-2; membrane-associated DHHC2 zinc finger protein; palmitoyltransferase ZDHHC2; Ream; rec; Reduced expression associated with metastasis protein; reduced expression in cancer protein; small rec; srec; testis tissue sperm-binding protein Li 56e; Zdhhc2; Zinc finger DHHC domain-containing protein 2; zinc finger DHHC-type containing 2; zinc finger protein 372; zinc finger, DHHC domain containing 2; zinc finger, DHHC-type containing 2; ZNF372
Gene Symbols ZDHHC2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 270-367 of human ZDHHC2 (NP_0574371).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246326, 51201
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.