Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZDHHC2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | ZDHHC2 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ZDHHC2 Polyclonal antibody specifically detects ZDHHC2 in Human samples. It is validated for ImmunofluorescenceSpecifications
ZDHHC2 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, GPCR, Signal Transduction | |
PBS, pH 7.2, 40% glycerol | |
51201 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
Human | |
DHHC2, DHHC-2, EC 2.3.1, EC 2.3.1.-, palmitoyltransferase ZDHHC2, Ream, REC, Reduced expression associated with metastasis protein, Reduced expression in cancer protein, Zinc finger DHHC domain-containing protein 2, Zinc finger protein 372, zinc finger, DHHC domain containing 2, zinc finger, DHHC-type containing 2, ZNF372rec | |
This antibody was developed against Recombinant Protein corresponding to amino acids: SSLGDGCSFPTCLVNQDPEQASTPAGLNSTAKNLENHQFPAKPLRESQSHLLTDSQSWTESSINPGKCKAGMSNPALTME | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title