Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZDHHC20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101778
Description
Antibody detects endogenous levels of total ZDHHC20.
Catalyzes palmitoylation of Cys residues on target proteins (PubMed:27153536, PubMed:29326245). Catalyzes palmitoylation of Cys residues in the cytoplasmic C-terminus of EGFR, and modulates the duration of EGFR signaling by modulating palmitoylation-dependent EGFR internalization and degradation (PubMed:27153536). Has a preference for acyl-CoA with C16 fatty acid chains (PubMed:29326245). Can also utilize acyl-CoA with C14 and C18 fatty acid chains (PubMed:29326245). [UniProt].
Specifications
ZDHHC20 | |
Polyclonal | |
Unconjugated | |
ZDHHC20 | |
4933421L13Rik; 5033406L14Rik; AI448102; B230110O18Rik; DHHC-20; DHHC-containing protein 20; HK11; membrane-associated DHHC24 zinc finger protein; Palmitoyltransferase ZDHHC20; probable palmitoyltransferase ZDHHC20; RGD1305755; ZDHHC20; Zinc finger DHHC domain-containing protein 20; zinc finger DHHC-type containing 20; zinc finger, DHHC domain containing 20; zinc finger, DHHC-type containing 20 | |
Rabbit | |
Affinity chromatography | |
RUO | |
253832, 305923, 75965 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5W0Z9, Q5Y5T1 | |
ZDHHC20 | |
A synthesized peptide derived from human ZDHHC20(Accession Q5W0Z9), corresponding to amino acid residues M71-A121. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction