Learn More
Invitrogen™ ZDHHC20 Polyclonal Antibody

Description
Positive Samples: HeLa, MCF7, SKOV3, A-549, Mouse brain, Mouse lung, Mouse testis, Rat brain, Rat lung Immunogen sequence: VFGDEKKYWL LPIFSSLGDG CSFPTRLVGM DPEQASVTNQ NEYARSSGSN QPFPIKPLSE SKNRLLDSES QWLENGAEEG IVKSGTNNHV TVAIEN.
Specifications
Specifications
| Antigen | ZDHHC20 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.54 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | ZDHHC20 |
| Gene Accession No. | Q5W0Z9, Q5Y5T1 |
| Gene Alias | 4933421L13Rik; 5033406L14Rik; AI448102; B230110O18Rik; DHHC-20; DHHC-containing protein 20; HK11; membrane-associated DHHC24 zinc finger protein; Palmitoyltransferase ZDHHC20; probable palmitoyltransferase ZDHHC20; RGD1305755; ZDHHC20; Zinc finger DHHC domain-containing protein 20; zinc finger DHHC-type containing 20; zinc finger, DHHC domain containing 20; zinc finger, DHHC-type containing 20 |
| Gene Symbols | ZDHHC20 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.