Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZDHHC20 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548558
Description

Specifications
ZDHHC20 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q5W0Z9 | |
ZDHHC20 | |
KLH conjugated synthetic peptide between 26-54 amino acids from the N-terminal region of human ZDHHC20. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
ZDHHC20 | |
Probable palmitoyltransferase ZDHHC20, Zinc finger DHHC domain-containing protein 20, DHHC-20, ZDHHC20 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
253832 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction