Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZDHHC21 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566757
Description
Immunogen sequence: RASITDPGRLP ENPKIPHGER EFWELCNKCN LMRPKRSHHC SRCGHCVRRM Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
Palmitoylates sex steroid hormone receptors, including ESR1, PGR and AR, thereby regulating their targeting to the plasma membrane. This affects rapid intracellular signaling by sex hormones via ERK and AKT kinases and the generation of cAMP, but does not affect that mediated by their nuclear receptor. [UniProt].
Specifications
ZDHHC21 | |
Polyclonal | |
Unconjugated | |
ZDHHC21 | |
9130404H11Rik; AL024349; D130004H04Rik; dep; DHHC21; DHHC-21; DNZ1; GABA-A receptor-associated membrane protein 3; Gramp3; HSPC097; palmitoyltransferase ZDHHC21; palmitoyltransferase ZDHHC21-like protein; probable palmitoyltransferase ZDHHC21; Zdhhc21; Zinc finger DHHC domain-containing protein 21; zinc finger DHHC-type containing 21; zinc finger DHHC-type palmitoyltransferase 21; zinc finger, DHHC domain containing 21; zinc finger, DHHC-type containing 21 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
340481 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IVQ6 | |
ZDHHC21 | |
Recombinant Human ZDHHC21. Recombinant protein control fragment (Product #RP-107414). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction