Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525096
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525096 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525096 Supplier Invitrogen™ Supplier No. PA525096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine based on sequence homology.

Palmitoylates sex steroid hormone receptors, including ESR1, PGR and AR, thereby regulating their targeting to the plasma membrane. This affects rapid intracellular signaling by sex hormones via ERK and AKT kinases and the generation of cAMP, but does not affect that mediated by their nuclear receptor. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC21
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ZDHHC21
Gene Accession No. Q8IVQ6, Q9D270
Gene Alias 9130404H11Rik; AL024349; D130004H04Rik; dep; DHHC21; DHHC-21; DNZ1; GABA-A receptor-associated membrane protein 3; Gramp3; HSPC097; palmitoyltransferase ZDHHC21; palmitoyltransferase ZDHHC21-like protein; probable palmitoyltransferase ZDHHC21; Zdhhc21; Zinc finger DHHC domain-containing protein 21; zinc finger DHHC-type containing 21; zinc finger DHHC-type palmitoyltransferase 21; zinc finger, DHHC domain containing 21; zinc finger, DHHC-type containing 21
Gene Symbols ZDHHC21
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 59-87 amino acids from the N-terminal region of human ZDHHC21.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340481, 68268
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.