Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC21 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985996
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5996 Supplier Bioss Supplier No. BS4229R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function of this protein has not been specifically defined.

Specifications

Antigen ZDHHC21
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZDHHC21
Gene Accession No. Q8IVQ6
Gene Alias 9130404H11Rik; AL024349; BOS_8751; D130004H04Rik; dep; DHHC21; DHHC-21; DNZ1; GABA-A receptor-associated membrane protein 3; Gramp3; HSPC097; LOC100062221; LOC100340626; Palmitoyltransferase ZDHHC21; Probable palmitoyltransferase ZDHHC21; probable palmitoyltransferase ZDHHC21-like; ZDHHC21; Zinc finger DHHC domain-containing protein 21; zinc finger DHHC-type containing 21; zinc finger, DHHC domain containing 21; zinc finger, DHHC domain containing 21-like; zinc finger, DHHC-type containing 21
Gene Symbols ZDHHC21
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZDHHC21.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340481
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.