Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572909
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572909 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572909 Supplier Invitrogen™ Supplier No. PA572909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZDHHC23 Polyclonal Antibody for Western Blot, IHC (P), IP

Palmitoyltransferase that mediates palmitoylation of KCNMA1, regulating localization of KCNMA1 to the plasma membrane. May be involved in NOS1 regulation and targeting to the synaptic membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC23
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.02% sodium azide
Gene ZDHHC23
Gene Accession No. Q8IYP9
Gene Alias DHHC-23; DHHC-containing protein 11; Gm1751; Gm779; NIDD; nNOS-intereacting DHHC-containing Dem protein-L; palmitoyltransferase ZDHHC23; probable palmitoyltransferase ZDHHC23; Zdhhc23; Zinc finger DHHC domain-containing protein 23; zinc finger DHHC-type containing 23; zinc finger, DHHC domain containing 23
Gene Symbols ZDHHC23
Host Species Rabbit
Immunogen Full-length recombinant human NIDD protein.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254887
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.