Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566681 Supplier Invitrogen™ Supplier No. PA566681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VYEFDEVMFPK NVRCSTCDLR KPARSK Highest antigen sequence identity to the following orthologs - mouse 85%, rat 96%.

ZDHHC4 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane; protein-cysteine S-palmitoyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZDHHC4
Gene Accession No. Q9NPG8
Gene Alias 1810021D01Rik; 2900029I10Rik; DC1; DHHC-4; membrane-associated DHHC4 zinc finger protein; probable palmitoyltransferase ZDHHC4; UNQ5787/PRO19576; Zdhhc4; Zinc finger DHHC domain-containing protein 4; zinc finger DHHC-type containing 4; zinc finger protein 374; zinc finger, DHHC domain containing 4; zinc finger, DHHC-type containing 4; ZNF374
Gene Symbols ZDHHC4
Host Species Rabbit
Immunogen Recombinant Human ZDHHC4. Recombinant protein control fragment (Product #RP-107330).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55146
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.