Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZDHHC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571404
Description
ZDHHC4 Polyclonal Antibody for Western Blot
ZDHHC4 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane; protein-cysteine S-palmitoyltransferase activity.
Specifications
ZDHHC4 | |
Polyclonal | |
Unconjugated | |
ZDHHC4 | |
1810021D01Rik; 2900029I10Rik; DC1; DHHC-4; membrane-associated DHHC4 zinc finger protein; probable palmitoyltransferase ZDHHC4; UNQ5787/PRO19576; Zdhhc4; Zinc finger DHHC domain-containing protein 4; zinc finger DHHC-type containing 4; zinc finger protein 374; zinc finger, DHHC domain containing 4; zinc finger, DHHC-type containing 4; ZNF374 | |
Rabbit | |
Affinity chromatography | |
RUO | |
304291 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5FVR1 | |
ZDHHC4 | |
synthetic peptide directed towards the C terminal region of rat ZDHHC4. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction