Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571409
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571409 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571409 Supplier Invitrogen™ Supplier No. PA571409
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZDHHC5 Polyclonal Antibody for Western Blot

Palmitoyl acyltransferase for the G-protein coupled receptor SSTR5. Also palmitoylates FLOT2. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Zdhhc5
Gene Accession No. Q9C0B5
Gene Alias 1110032A17Rik; AI451382; DHHC5; DHHC-5; Kiaa1748; membrane-associated DHHC5 zinc finger protein; palmitoyltransferase ZDHHC5; probable palmitoyltransferase ZDHHC5; ZDHHC5; Zinc finger DHHC domain-containing protein 5; zinc finger DHHC-type containing 5; zinc finger protein 375; zinc finger, DHHC domain containing 5; zinc finger, DHHC-type containing 5; Zisp; ZNF375
Gene Symbols Zdhhc5
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ZDHHC5.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25921
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.