Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553209
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553209 Supplier Invitrogen™ Supplier No. PA553209
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DPPLGYTSPF LSARLAQQRE AERHPRLVPT GPTHREPSPV RYDNLSRHIV ASLQEREKLL RQSPPLPGRE EEPGLGDSGI QSTPGSGHAP RTSSSSDDSK RSPLGKTPLG RPAVPRFGKP DGLRGRGVGS PE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.

Palmitoyl acyltransferase for the G-protein coupled receptor SSTR5. Also palmitoylates FLOT2. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Zdhhc5
Gene Accession No. Q9C0B5
Gene Alias 1110032A17Rik; AI451382; DHHC5; DHHC-5; Kiaa1748; membrane-associated DHHC5 zinc finger protein; palmitoyltransferase ZDHHC5; probable palmitoyltransferase ZDHHC5; ZDHHC5; Zinc finger DHHC domain-containing protein 5; zinc finger DHHC-type containing 5; zinc finger protein 375; zinc finger, DHHC domain containing 5; zinc finger, DHHC-type containing 5; Zisp; ZNF375
Gene Symbols Zdhhc5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZDHHC5. Recombinant protein control fragment (Product #RP-90998).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25921
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.