Learn More
ZDHHC5 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304653AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Orangutan and Chimpanzee
Palmitoyl acyltransferase for the G-protein coupled receptor SSTR5 and for FLOT2.Specifications
ZDHHC5 | |
Polyclonal | |
Unconjugated | |
ZDHHC5 | |
DHHC5, DHHC-5, KIAA1748, membrane-associated DHHC5 zinc finger protein, probable palmitoyltransferase ZDHHC5, zinc finger DHHC domain-containing protein 5, zinc finger protein 375, zinc finger, DHHC domain containing 5, zinc finger, DHHC-type containing 5, ZNF375 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25921 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9C0B5 | |
ZDHHC5 | |
Between 600 and 650 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.