Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC5 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501568299
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8299 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8299 Supplier Bethyl Laboratories, Inc Supplier No. A304652A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Dog, Orangutan and Chimpanzee

Palmitoyl acyltransferase for the G-protein coupled receptor SSTR5 and for FLOT2.

Specifications

Antigen ZDHHC5
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ZDHHC5
Gene Accession No. Q8VDZ4, Q9C0B5
Gene Alias DHHC5, DHHC-5, KIAA1748, membrane-associated DHHC5 zinc finger protein, probable palmitoyltransferase ZDHHC5, zinc finger DHHC domain-containing protein 5, zinc finger protein 375, zinc finger, DHHC domain containing 5, zinc finger, DHHC-type containing 5, ZNF375
Gene Symbols ZDHHC5
Host Species Rabbit
Immunogen Between 500 and 550
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228136, 25921
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.