Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZDHHC6 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548717
Description
Predicted to react with bovine based on sequence homology.
Palmitoylates calnexin (CALX), which is required for its association with the ribosome-translocon complex and efficient folding of glycosylated proteins.
Specifications
ZDHHC6 | |
Polyclonal | |
Unconjugated | |
ZDHHC6 | |
Palmitoyltransferase ZDHHC6, Transmembrane protein H4, Zinc finger DHHC domain-containing protein 6, DHHC-6, Zinc finger protein 376, ZDHHC6, ZNF376 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
64429, 66980 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide | |
Q9H6R6, Q9CPV7 | |
ZDHHC6 | |
KLH conjugated synthetic peptide between 288-317 amino acids from the C-terminal region of human ZDHHC6. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction