Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC7 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA563760

Catalog No. PIPA563760


Only null left
Add to Cart

Description

Description

Immunogen sequence: MQPSGHRLRD VEHHPLLAEN DNYDSSSSS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

ZDHHC7 is a protein coding gene. Palmitoyltransferase with broad specificity. Palmitoylates JAM3. Palmitoylates SNAP25 and DLG4/PSD95. Palmitoylates sex steroid hormone receptors, including ESR1, PGR and AR, thereby regulating their targeting to the plasma membrane and their function in rapid intracellular signaling upon binding of sex hormones. May play a role in follicle stimulation hormone (FSH) activation of testicular Sertoli cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZDHHC7
Polyclonal
Unconjugated
ZDHHC7
AL024087; DHHC7; DHHC-7; GABA-A receptor-associated membrane protein 2; Gramp2; membrane-associated DHHC7 zinc finger protein; palmitoyltransferase ZDHHC7; putative zinc finger protein SERZ-1; sertoli cell gene with a zinc finger domain protein; Sertoli cell gene with zinc finger domain-β Serz; SERZ1; Serz-1; SERZ-B; Zdhhc7; Zinc finger DHHC domain-containing protein 7; zinc finger DHHC-type containing 7; zinc finger protein 370; zinc finger, DHHC domain containing 7; zinc finger, DHHC-type containing 7; ZNF370
Rabbit
Antigen affinity chromatography
RUO
55625
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NXF8
ZDHHC7
Recombinant protein corresponding to Human ZDHHC7. Recombinant protein control fragment (Product #RP-101935).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.