Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584399
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584399 Supplier Invitrogen™ Supplier No. PA584399
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKSENPTWER RLRWEGMKSV FGGPPSLLWM NPFVGFRFRR LPTRPRKGGP EFS Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 91%.

ZDHHC7 is a protein coding gene. Palmitoyltransferase with broad specificity. Palmitoylates JAM3. Palmitoylates SNAP25 and DLG4/PSD95. Palmitoylates sex steroid hormone receptors, including ESR1, PGR and AR, thereby regulating their targeting to the plasma membrane and their function in rapid intracellular signaling upon binding of sex hormones. May play a role in follicle stimulation hormone (FSH) activation of testicular Sertoli cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC7
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZDHHC7
Gene Accession No. Q9NXF8
Gene Alias AL024087; DHHC7; DHHC-7; GABA-A receptor-associated membrane protein 2; Gramp2; membrane-associated DHHC7 zinc finger protein; palmitoyltransferase ZDHHC7; putative zinc finger protein SERZ-1; sertoli cell gene with a zinc finger domain protein; Sertoli cell gene with zinc finger domain-β Serz; SERZ1; Serz-1; SERZ-B; Zdhhc7; Zinc finger DHHC domain-containing protein 7; zinc finger DHHC-type containing 7; zinc finger protein 370; zinc finger, DHHC domain containing 7; zinc finger, DHHC-type containing 7; ZNF370
Gene Symbols ZDHHC7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZDHHC7. Recombinant protein control fragment (Product #RP-106910).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55625
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.