Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ ZDHHC8 Polyclonal Antibody
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518664
Description
This antibody is predicted to react with canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a four transmembrane protein that is a member of the zinc finger DHHC domain-containing protein family. The encoded protein may function as a palmitoyltransferase. Defects in this gene may be associated with a susceptibility to schizophrenia. Alternate splicing of this gene results in multiple transcript variants. A pseudogene of this gene is found on chromosome 22. 
            Specifications
| ZDHHC8 | |
| Polyclonal | |
| Unconjugated | |
| Zdhhc8 | |
| D16H22S1738E; DHHC8; DHHC-8; DHHC-containing protein 8; E330009O14Rik; KIAA1292; membrane-associated DHHC8 zinc finger protein; Op53c05; Probable palmitoyltransferase ZDHHC8; ZDHHC8; ZDHHCL1; zinc finger DHHC domain-containing protein 8; zinc finger DHHC-type containing 8; zinc finger protein 378; zinc finger, DHHC domain containing 8; zinc finger, DHHC domain like containing 1; zinc finger, DHHC-type containing 8; zinc finger, DHHC-type containing 8-like; ZNF378 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 29801 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | 
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9ULC8 | |
| Zdhhc8 | |
| Synthetic peptide sequence (QRDHPQLKTPPSK) corresponding to the internal amino acids of ZDHHC8 (aa 708-721). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            