Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551931 Supplier Invitrogen™ Supplier No. PA551931
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKIEAGTFSS DLQTPRPGSA ESALSVQRTS PPTPAMYKFR PAFPTGPKVP FCGPGEQVPG PDSLTLGDDS IRSLDFVSEP SLDLPDYGPG GLHAAYPPSP PLSASDAFSG ALRSLSLKAS SRRGGDHVAL Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.

This gene encodes a four transmembrane protein that is a member of the zinc finger DHHC domain-containing protein family. The encoded protein may function as a palmitoyltransferase. Defects in this gene may be associated with a susceptibility to schizophrenia. Alternate splicing of this gene results in multiple transcript variants. A pseudogene of this gene is found on chromosome 22.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC8
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Zdhhc8
Gene Accession No. Q9ULC8
Gene Alias D16H22S1738E; DHHC8; DHHC-8; DHHC-containing protein 8; E330009O14Rik; KIAA1292; membrane-associated DHHC8 zinc finger protein; Op53c05; Probable palmitoyltransferase ZDHHC8; ZDHHC8; ZDHHCL1; zinc finger DHHC domain-containing protein 8; zinc finger DHHC-type containing 8; zinc finger protein 378; zinc finger, DHHC domain containing 8; zinc finger, DHHC domain like containing 1; zinc finger, DHHC-type containing 8; zinc finger, DHHC-type containing 8-like; ZNF378
Gene Symbols Zdhhc8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZDHHC8. Recombinant protein control fragment (Product #RP-91540).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29801
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.