Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110591 Supplier Invitrogen™ Supplier No. PA5110591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VEHVLCSPLA PRYVVEPPRL PLAVSLKPPF LRPELLDRAA PLKVKLSDNG LKAGLGRSKS KGSLDRLDEK PLDLGPPLPP KIEAGTFSSD LQTPRPGSAE SALSVQRTSP PTPAMYKFRP AFPTGPKVPF CGPGEQVPGP D.

This gene encodes a four transmembrane protein that is a member of the zinc finger DHHC domain-containing protein family. The encoded protein may function as a palmitoyltransferase. Defects in this gene may be associated with a susceptibility to schizophrenia. Alternate splicing of this gene results in multiple transcript variants. A pseudogene of this gene is found on chromosome 22.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Zdhhc8
Gene Accession No. Q5Y5T5, Q9ULC8
Gene Alias D16H22S1738E; DHHC8; DHHC-8; DHHC-containing protein 8; E330009O14Rik; KIAA1292; membrane-associated DHHC8 zinc finger protein; Op53c05; Probable palmitoyltransferase ZDHHC8; ZDHHC8; ZDHHCL1; zinc finger DHHC domain-containing protein 8; zinc finger DHHC-type containing 8; zinc finger protein 378; zinc finger, DHHC domain containing 8; zinc finger, DHHC domain like containing 1; zinc finger, DHHC-type containing 8; zinc finger, DHHC-type containing 8-like; ZNF378
Gene Symbols Zdhhc8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 240-380 of human ZDHHC8 (NP_037505.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27801, 29801
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.