Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZDHHC8 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS18486R

Description
This gene encodes a four transmembrane protein that is a member of the zinc finger DHHC domain-containing protein family. The encoded protein may function as a palmitoyltransferase. Defects in this gene may be associated with a susceptibility to schizophrenia. Alternate splicing of this gene results in multiple transcript variants. A pseudogene of this gene is found on chromosome 22.Specifications
ZDHHC8 | |
Polyclonal | |
Unconjugated | |
Zdhhc8 | |
D16H22S1738E; DHHC8; DHHC-8; DHHC-containing protein 8; E330009O14Rik; KIAA1292; membrane-associated DHHC8 zinc finger protein; Op53c05; Probable palmitoyltransferase ZDHHC8; ZDHHC8; ZDHHCL1; Zinc finger DHHC domain-containing protein 8; zinc finger DHHC-type containing 8; Zinc finger protein 378; zinc finger, DHHC domain containing 8; zinc finger, DHHC domain like containing 1; zinc finger, DHHC-type containing 8; ZNF378 | |
Rabbit | |
Protein A | |
RUO | |
29801 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9ULC8 | |
Zdhhc8 | |
KLH conjugated synthetic peptide derived from human ZDHHC8/ZNF378. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction