Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556868
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556868 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556868 Supplier Invitrogen™ Supplier No. PA556868
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSTQETSSSL LPQSPAPTEH LNSNEMPEDS STPEEMPPPE PPEPPQEAAE AEK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.

The ZDHHC9-GOLGA7 complex is a palmitoyltransferase specific for HRAS and NRAS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZDHHC9
Gene Accession No. Q9Y397
Gene Alias 6430508G22; 9530098M12Rik; antigen MMSA-1; Asp-His-His-Cys domain containing protein 9; CGI89; CGI-89; CXorf11; DHHC9; DHHC-9; membrane-associated DHHC9 zinc finger protein; MMSA1; MRXSZ; Palmitoyltransferase ZDHHC9; UNQ261/PRO298; ZDHHC10; ZDHHC9; Zinc finger DHHC domain-containing protein 9; zinc finger DHHC-type containing 9; zinc finger protein 379; Zinc finger protein 380; zinc finger, DHHC domain containing 10; zinc finger, DHHC domain containing 9; zinc finger, DHHC-type containing 9; ZNF379; ZNF380
Gene Symbols ZDHHC9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZDHHC9. Recombinant protein control fragment (Product #RP-95472).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51114
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.