Learn More
Invitrogen™ ZDHHC9 Polyclonal Antibody

Description
Immunogen sequence: VALNQTTNED IKGSWTGKNR VQNPYSHGNI VKNCCEVLCG PLPPSVLDRR GILPLEESGS RPPSTQETSS SLLPQSPAPT EHLNSNEMPE DSSTPEEMPP PEPPEPPQEA AEAEK.
Specifications
Specifications
| Antigen | ZDHHC9 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ZDHHC9 |
| Gene Accession No. | P59268, Q9Y397 |
| Gene Alias | 6430508G22; 9530098M12Rik; antigen MMSA-1; Asp-His-His-Cys domain containing protein 9; CGI89; CGI-89; CXorf11; DHHC9; DHHC-9; membrane-associated DHHC9 zinc finger protein; MMSA1; MRXSZ; Palmitoyltransferase ZDHHC9; UNQ261/PRO298; ZDHHC10; ZDHHC9; Zinc finger DHHC domain-containing protein 9; zinc finger DHHC-type containing 9; zinc finger protein 379; Zinc finger protein 380; zinc finger, DHHC domain containing 10; zinc finger, DHHC domain containing 9; zinc finger, DHHC-type containing 9; ZNF379; ZNF380 |
| Gene Symbols | ZDHHC9 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.