Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZDHHC9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ZDHHC9 |
---|---|
Concentration | 0.34 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4218
![]() ![]() |
Proteintech Group Inc
240461AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The ZDHHC9-GOLGA7 complex is a palmitoyltransferase specific for HRAS and NRAS.Specifications
ZDHHC9 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P59268, Q9Y397 | |
208884, 51114 | |
ZDHHC9 Fusion Protein Ag20881 | |
Primary | |
-20°C | |
ZDHHC9 |
0.34 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CGI 89, CXorf11, DHHC 9, DHHC9, MMSA 1, Palmitoyltransferase ZDHHC9, UNQ261/PRO298, ZDHHC10, ZDHHC9, Zinc finger protein 379, ZNF379, ZNF380 | |
ZDHHC9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title