Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC9 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502209696
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9696 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9696 Supplier Fabgennix Inc Supplier No. ZDHHC9901AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZDHHC9 Polyclonal antibody specifically detects ZDHHC9 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen ZDHHC9
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene ZDHHC9
Gene Accession No. P59268, Q9Y397
Gene Alias 6430508G22; 9530098M12Rik; antigen MMSA-1; Asp-His-His-Cys domain containing protein 9; CGI89; CGI-89; CXorf11; DHHC9; DHHC-9; membrane-associated DHHC9 zinc finger protein; MMSA1; MRXSZ; Palmitoyltransferase ZDHHC9; UNQ261/PRO298; ZDHHC10; ZDHHC9; Zinc finger DHHC domain-containing protein 9; zinc finger DHHC-type containing 9; zinc finger protein 379; Zinc finger protein 380; zinc finger, DHHC domain containing 10; zinc finger, DHHC domain containing 9; zinc finger, DHHC-type containing 9; ZNF379; ZNF380
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 250-300 on human Palmitoyltransferase ZDHHC9 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208884, 302808, 51114
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.