Learn More
Invitrogen™ Zearalenone Monoclonal Antibody (6D3)

Description
Antibody tested in indirect ELISA.
Specifications
Specifications
| Antigen | Zearalenone |
| Applications | ELISA, Immunohistochemistry, Lateral Flow, SDS-Page |
| Classification | Monoclonal |
| Clone | 6D3 |
| Concentration | 10.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Host Species | Mouse |
| Immunogen | Conjugate of ZEN hapten and KLH. |
| Purification Method | Protein G |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.