Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZEB1 Antibody (2A8A6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications
$206.50 - $499.50
Specifications
Antigen | ZEB1 |
---|---|
Clone | 2A8A6 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1: 500 - 1:2000. Use reported by customer review and in scientific literature (PMID 29383119), Flow Cytometry 1:200 - 1:400, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP223484SS
![]() |
Novus Biologicals
NBP223484SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP223484
![]() |
Novus Biologicals
NBP223484 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
ZEB1 Monoclonal specifically detects ZEB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ZEB1 | |
1.0 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Chromatin Research, Immunology, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision, Zinc Finger | |
P37275 | |
6935 | |
Partial recombinant human ZEB1 (between residues 800-1124) expressed in E. coli [Uniprot P37275] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
2A8A6 | |
Western Blot 1: 500 - 1:2000. Use reported by customer review and in scientific literature (PMID 29383119), Flow Cytometry 1:200 - 1:400, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
Monoclonal | |
Precipitation | |
RUO | |
Human, Rat | |
AREB6MGC133261, delta-crystallin enhancer binding factor 1, DELTAEF1, FECD6, Negative regulator of IL2, NIL2A, NIL-2-A, NIL-2-A zinc finger protein, posterior polymorphous corneal dystrophy 3, PPCD3, TCF-8, TCF8BZP, Transcription factor 8, transcription factor 8 (represses interleukin 2 expression), ZEB, Zfhep, ZFHX1A, zinc finger E-box binding homeobox 1, zinc finger E-box-binding homeobox 1, zinc finger homeodomain enhancer-binding protein | |
ZEB1 | |
IgG1 | |
Protein A or G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title