Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZEB1 Monoclonal Antibody (OTI7E12), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA802313

Description
This gene encodes a zinc finger transcription factor. The encoded protein likely plays a role in transcriptional repression of interleukin 2. Mutations in this gene have been associated with posterior polymorphous corneal dystrophy-3 and late-onset Fuchs endothelial corneal dystrophy. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
ZEB1 | |
Monoclonal | |
Unconjugated | |
ZEB1 | |
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A | |
Mouse | |
Affinity Chromatography | |
RUO | |
6935 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7E12 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P37275 | |
ZEB1 | |
Human recombit protein fragment corresponding to amino acids 1-350 of human ZEB1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction