Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZEB1 Antibody (2A8A6) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281015
Description
ZEB1 Monoclonal specifically detects ZEB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
ZEB1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1: 500 - 1:2000. Use reported by customer review and in scientific literature (PMID 29383119), Flow Cytometry : 1:200 - 1:400, Immunohistochemistry : 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence : 1:100, Immunohistochemistry-Paraffin : 1:200 - 1:1000, CyTOF-ready | |
AREB6MGC133261, delta-crystallin enhancer binding factor 1, DELTAEF1, FECD6, Negative regulator of IL2, NIL2A, NIL-2-A, NIL-2-A zinc finger protein, posterior polymorphous corneal dystrophy 3, PPCD3, TCF-8, TCF8BZP, Transcription factor 8, transcription factor 8 (represses interleukin 2 expression), ZEB, Zfhep, ZFHX1A, zinc finger E-box binding homeobox 1, zinc finger E-box-binding homeobox 1, zinc finger homeodomain enhancer-binding protein | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
2A8A6 | |
Unconjugated | |
PBS | |
ZEB1 | |
Partial recombinant human ZEB1 (between residues 800-1124) expressed in E. coli [Uniprot P37275] | |
0.1 mL | |
Cancer, Chromatin Research, Immunology, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision, Zinc Finger | |
6935 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction