Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZEB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540350
Description
Peptide sequence: NPRRNNVTNY NTVVETNSDS DDEDKLHIVE EESVTDAADC EGVPEDDLPT Sequence homology: Cow: 100%; Dog: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%; Zebrafish: 100%.
ZEB1 (Zinc finger E-box-binding homeobox 1) acts as a transcriptional repressor. It inhibits interleukin-2 (IL-2) gene expression. ZEB1 enhances or represses the promoter activity of the ATP1A1 gene depending on the quantity of cDNA and depending on the cell type. It represses E-cadherin promoters and induces an epithelial-mesenchymal transition (EMT) by recruiting SMARCA4/BRG1. ZEB1 also represses BCL6 transcription in the presence of the corepressor CTBP1. It postively regulates neuronal differentiation and represses RCOR1 transcription activation during neurogenesis. Mutations in the gene can result in corneal dystrophy, posterior polymorphous and corneal dystrophy, Fuchs endothelial 6.
Specifications
ZEB1 | |
Polyclonal | |
Unconjugated | |
ZEB1 | |
[delta]EF1; 3110032K11Rik; AREB6; BZP; Delta EF1; delta-crystallin enhancer binding factor 1; deltaEF1; DELTA-EF1; FECD6; LOW QUALITY PROTEIN: zinc finger E-box-binding homeobox 1; MEB1; MGC133261; Negative regulator of IL2; Nil2; NIL2A; NIL-2A; NIL-2-A; NIL-2-A zinc finger protein; OTTHUMP00000019405; posterior polymorphous corneal dystrophy 3; PPCD3; RP11-472N13.4; Tcf18; TCF8; TCF-8; Transcription factor 8; transcription factor 8 (represses interleukin 2 expression); Tw; twirler; ZEB; Zeb1; Zfhep; Zfhx1a; Zfx1a; Zfx1ha; zinc finger E-box binding homeobox 1; zinc finger E-box-binding homeobox 1; Zinc finger homeobox protein 1a; zinc finger homeodomain enhancer-binding protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6935 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P37275 | |
ZEB1 | |
Synthetic peptide directed towards the N-terminal of human ZEB1 (aa 14-63). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction