Learn More
ZEB1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301922A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
This gene encodes a zinc finger transcription factor. The encoded protein likely plays a role in transcriptional repression of interleukin 2. Mutations in this gene have been associated with posterior polymorphous corneal dystrophy-3 and late-onset Fuchs endothelial corneal dystrophy. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
ZEB1 | |
Polyclonal | |
Unconjugated | |
ZEB1 | |
ZEB1; AREB6; zinc finger E-box binding homeobox 1; BZP; DELTA-EF1; MGC133261; NIL-2-A; NIL-2A; NIL2A; TCF8; ZEB; ZFHEP; ZFHX1A; OTTHUMP00000019405; transcription factor 8 (represses interleukin 2 expression); zinc finger homeodomain enhancer-binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21417, 6935 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P37275, Q64318 | |
ZEB1 | |
Between 1074 and 1124 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.