Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZEB1 Recombinant Rabbit Monoclonal Antibody (BLR102H)
GREENER_CHOICE

Catalog No. PIMA544395
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544395 100 μL
PIMA544396 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544395 Supplier Invitrogen™ Supplier No. MA544395
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

ZEB1 Recombinant Monoclonal Antibody for Western Blot, IP

ZEB1 (Zinc finger E-box-binding homeobox 1) acts as a transcriptional repressor. It inhibits interleukin-2 (IL-2) gene expression. ZEB1 enhances or represses the promoter activity of the ATP1A1 gene depending on the quantity of cDNA and depending on the cell type. It represses E-cadherin promoters and induces an epithelial-mesenchymal transition (EMT) by recruiting SMARCA4/BRG1. ZEB1 also represses BCL6 transcription in the presence of the corepressor CTBP1. It postively regulates neuronal differentiation and represses RCOR1 transcription activation during neurogenesis. Mutations in the gene can result in corneal dystrophy, posterior polymorphous and corneal dystrophy, Fuchs endothelial 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZEB1
Applications Immunoprecipitation, Western Blot, Western Blot
Classification Recombinant Monoclonal
Clone BLR102H
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6
Gene ZEB1
Gene Accession No. P37275, Q64318
Gene Alias [delta]EF1; 3110032K11Rik; AREB6; BZP; Delta EF1; delta-crystallin enhancer binding factor 1; deltaEF1; DELTA-EF1; FECD6; LOW QUALITY PROTEIN: zinc finger E-box-binding homeobox 1; MEB1; MGC133261; Negative regulator of IL2; Nil2; NIL2A; NIL-2A; NIL-2-A; NIL-2-A zinc finger protein; OTTHUMP00000019405; posterior polymorphous corneal dystrophy 3; PPCD3; RP11-472N13.4; Tcf18; TCF8; TCF-8; Transcription factor 8; transcription factor 8 (represses interleukin 2 expression); Tw; twirler; ZEB; Zeb1; Zfhep; Zfhx1a; Zfx1a; Zfx1ha; zinc finger E-box binding homeobox 1; zinc finger E-box-binding homeobox 1; Zinc finger homeobox protein 1a; zinc finger homeodomain enhancer-binding protein
Gene Symbols ZEB1
Host Species Rabbit
Immunogen Peptide representing a region between residue 1074 and C-terminus (residue 1124) of human zinc finger E-box-binding homeobox 1.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21417, 6935
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.