Learn More
Invitrogen™ ZEB1 Recombinant Rabbit Monoclonal Antibody (BLR102H)
Description
ZEB1 Recombinant Monoclonal Antibody for Western Blot, IP
Specifications
Specifications
| Antigen | ZEB1 |
| Applications | Immunoprecipitation, Western Blot, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | BLR102H |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 |
| Gene | ZEB1 |
| Gene Accession No. | P37275, Q64318 |
| Gene Alias | [delta]EF1; 3110032K11Rik; AREB6; BZP; Delta EF1; delta-crystallin enhancer binding factor 1; deltaEF1; DELTA-EF1; FECD6; LOW QUALITY PROTEIN: zinc finger E-box-binding homeobox 1; MEB1; MGC133261; Negative regulator of IL2; Nil2; NIL2A; NIL-2A; NIL-2-A; NIL-2-A zinc finger protein; OTTHUMP00000019405; posterior polymorphous corneal dystrophy 3; PPCD3; RP11-472N13.4; Tcf18; TCF8; TCF-8; Transcription factor 8; transcription factor 8 (represses interleukin 2 expression); Tw; twirler; ZEB; Zeb1; Zfhep; Zfhx1a; Zfx1a; Zfx1ha; zinc finger E-box binding homeobox 1; zinc finger E-box-binding homeobox 1; Zinc finger homeobox protein 1a; zinc finger homeodomain enhancer-binding protein |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.