Learn More
Thermo Scientific™ Zeba™ Dye and Biotin Removal Spin Columns, 2 mL, 5 Columns PROMO
Description
Removing free-dye after a labeling reaction is often difficult and time-consuming but is essential for accurate determination of dye-to-protein ratios. Pierce Zeba Dye and Biotin Removal resin is highly specialized to produce exceptional protein recoveries while effectively removing non-conjugated fluorescent dyes, biotinylation reagents, reducing agents, and crosslinkers. When using the appropriate amount of sample and working within buffer conditions, these columns and filter plates can successfully remove almost any fluorescent dye. Even dilute protein samples (25 μg/mL) can be successfully processed to remove dyes, biotin, reducing agents, and crosslinkers and to efficiently recover proteins (>7 kDa). These products are not recommended for desalting or buffer exchange.
Features of Zeba Dye and Biotin Removal columns and filter plates include:
- Versatile—removes unreacted fluorescent dyes, biotinylation reagents, crosslinkers, and reducing agents from protein solutions
- High recovery—low-binding resin maximizes protein recovery
- Easy-to-use—no cumbersome column preparation or equilibration
- Fast—small-molecule removal and protein recovery in less than 15 minutes
- Flexible—available in spin columns and filter spin plates for a range of needs
Sample capacities
- 0.5-mL spin column: 50 to 120 μL sample
- 2-mL spin column: 400 to 700 μL sample
- 5-mL spin column: 1 to 2 mL sample
- 96-well filter plate: 40 to 100 μL sample (per well)
Available formats
- Spin columns: 0.5, 2, 5, and 10 mL
- 96-well filter plates: pre-dispensed; compatible with centrifugation for manual or automated purification; enable fast, consistent well-to-well and plate-to-plate reproducibility for small-scale, high-throughput clean-ups
Applications
- Ideal for post-reaction clean-up of samples
- To effectively remove non-conjugated dye, biotin, reducing agents, and crosslinkers with exceptional protein recovery
Specifications
Specifications
Content And Storage | Store in Refrigerator (2°C to 8°C). |
Shipping Condition | Room Temperature |
Format | Spin Column |
Column Type | Proprietary Resin |
Product Line | Zeba™ |
Quantity | 5 Columns |
Volume (Metric) | 2 mL |
Sample Volume (Metric) | 400 to 700 μL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.