Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zebrafish Basement Membrane Marker Antibody (FIS 5F11/2), Janelia Fluor™ 549, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP250369JF549

Catalog No. NBP50369549


Only null left
Add to Cart

Description

Description

Zebrafish Basement Membrane Marker Monoclonal specifically detects Zebrafish Basement Membrane Marker in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ZebrafishBasementMembraneMarker
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
Zebrafish Intestine
0.1 mL
Primary
Store at 4°C in the dark.
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
FIS 5F11/2
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Mouse
Protein A purified
Developmental Biology, Epitope Tags
Zebrafish
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.