Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zebrafish Delta C Antibody (zdc2), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250371UV
Description
Zebrafish Delta C Monoclonal specifically detects Zebrafish Delta C in Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.Specifications
Zebrafish Delta C | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Extracellular region of Zebrafish Delta C protein | |
0.1 mL | |
Cell Cycle and Replication, Developmental Biology, Neuroscience | |
Zebrafish | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
zdc2 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction