Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zebrafish Delta C Antibody (zdc2), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250371APCCY7
Description
Zebrafish Delta C Monoclonal specifically detects Zebrafish Delta C in Zebrafish samples. It is validated for Flow Cytometry.Specifications
Zebrafish Delta C | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
Extracellular region of Zebrafish Delta C protein | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
zdc2 | |
Flow Cytometry | |
Mouse | |
Protein A purified | |
Cell Cycle and Replication, Developmental Biology, Neuroscience | |
Zebrafish | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction