Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zebrafish Delta D Antibody (zdd2), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP50372646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP50372646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP50372646 Supplier Novus Biologicals Supplier No. NBP250372JF646
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Zebrafish Delta D Monoclonal specifically detects Zebrafish Delta D in Zebrafish samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ZebrafishDeltaD
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone zdd2
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Extracellular domain; Zebrafish Delta D peptide
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Developmental Biology, Neuroscience
Primary or Secondary Primary
Test Specificity Specific for Zebrafish Delta D extracellular domain. This antibody does not detect Delta C, but it cross-reacts weakly with Delta A.
Target Species Zebrafish
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.