Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zebrafish Delta D Antibody (zdd2), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250372MFV450
Description
Zebrafish Delta D Monoclonal antibody specifically detects Zebrafish Delta D in Zebrafish samples. It is validated for Flow CytometrySpecifications
Zebrafish Delta D | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Extracellular domain; Zebrafish Delta D peptide | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
zdd2 | |
Flow Cytometry | |
Mouse | |
Protein A purified | |
RUO | |
Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction