Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zebrafish Ephrin-B2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Ephrin-B2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Ephrin-B2 Polyclonal specifically detects Ephrin-B2 in Zebrafish samples. It is validated for Western Blot.Specifications
Ephrin-B2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Zebrafish | |
EFNB2, ELF-2, ephrin B2, EphrinB2, EPLG5, Htk-L, HTKL, LERK5, LERK-5, NLERK-1 | |
efnb2a | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
O73874 | |
1948 | |
Mouse myeloma cell line NS0-derived recombinant zebrafish Ephrin-B2 Leu25-Ala222 Accession # O73874 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects zebrafish Ephrin-B2 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) Ephrin-B2 is observed and less than 1% cross-reactivity with recombinant human (rh) Ephrin-A4, rmEphrin-A2, rmEphrin-A1, rhEphrin-B3, rmEphrin-B1, rhEphrin-A3, rhEphrin-A5, and rmEphrin-A4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title