Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zebrafish Gut Secretory Cell Epitopes Antibody (FIS 2F11/2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2503750.025ML
Description
Zebrafish Gut Secretory Cell Epitopes Monoclonal antibody specifically detects Zebrafish Gut Secretory Cell Epitopes in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| Zebrafish Gut Secretory Cell Epitopes | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| FIS 2F11/2 | |
| Unconjugated | |
| PBS | |
| Lysate of zebrafish intestine | |
| 0.025 mL | |
| Cell Cycle and Replication, Cellular Signaling, Developmental Biology, Epigenetics, Epitope Tags | |
| Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction