Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zebrafish Gut Secretory Cell Epitopes Antibody (FIS 2F11/2), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250375R
Description
Zebrafish Gut Secretory Cell Epitopes Monoclonal specifically detects Zebrafish Gut Secretory Cell Epitopes in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Zebrafish Gut Secretory Cell Epitopes | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Lysate of zebrafish intestine | |
0.1 mL | |
Cell Cycle and Replication, Cellular Signaling, Developmental Biology, Epigenetics, Epitope Tags | |
Zebrafish | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
FIS 2F11/2 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction