Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zebrafish Gut Secretory Cell Epitopes Antibody (FIS 4B7/2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Zebrafish Gut Secretory Cell Epitopes |
---|---|
Clone | FIS 4B7/2 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:200, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Description
Zebrafish Gut Secretory Cell Epitopes Monoclonal antibody specifically detects Zebrafish Gut Secretory Cell Epitopes in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
Zebrafish Gut Secretory Cell Epitopes | |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication, Cellular Signaling, Developmental Biology, Epigenetics, Epitope Tags | |
PBS | |
IgG1 | |
Protein A purified |
FIS 4B7/2 | |
Western Blot 1:200, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Zebrafish | |
Lysate of fish intestine | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title