Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zebrafish VEGF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1247
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1247 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1247 Supplier R&D Systems Supplier No. MAB1247
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

VEGF Monoclonal specifically detects VEGF in Zebrafish samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen VEGF
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 211615
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.2-0.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O73682.1
Gene Alias MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Gene Symbols vegfaa
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant zebrafish VEGF 165 Ala24-Arg188 Accession # O73682.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7422
Test Specificity Detects zebrafish VEGF in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) VEGF 121 165 206 rhVEGF-B 186 rhVEGF-C, rhVEGF-D, recombinant mouse (rm) VEGF 115 rmVEGF 165 rmVEGF-B 186 rmVEGF-D, or recombinant rat VEGF 164 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Zebrafish
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.