Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Zebularine
SDP

Catalog No. 229310 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-931-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-931-0 Supplier R&D Systems™ Supplier No. 2293/10

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. DNA methyltransferase and cytidine deaminase inhibitor

Orally active DNA methyltransferase inhibitor. Inhibits tumor cell proliferation (IC50 = 120 μM) and re-activates silenced genes in T24 bladder carcinoma cells. Inhibits cytidine deaminase (Ki ∽ 2 μM); induces differentiation of mesenchymal stem cells into cardiomyocytes.

Chemical Identifiers

CAS 3724-10-5
Molecular Formula C8H8O2S

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 228.2
Purity >99%
Molecular Weight (g/mol) 168.21
Chemical Name or Material Zebularine
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.