Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zenker's Fixative Solution, Ricca Chemical
SDP

Catalog No. p-3252632 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Mercuric Chloride-Potassium Dichromate-Sodium Sulfate Mixture

Specifications

Physical Form Liquid
Chemical Name or Material Zenker's Fixative Solution
Name Note Mercuric Chloride-Potassium Dichromate-Sodium Sulfate Mixture
CAS 7732-18-5, 7487-94-7, 7778-50-9, 7757-82-6
CAS Min % 90.0, 4.52, 2.26, 0.9
CAS Max % 94.0, 4.7, 2.35, 0.94
Packaging Natural Poly Bottle
Color Orange
Quantity 1 L
Grade Laboratory
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.