Alfa Aesar™ Zeolite ZSM-5, ammonium

Manufacturer: Alfa Aesar™ 4588014

Quantity 25g

 View more versions of this product

Catalog No. AA4588014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1318-02-1
    MDL Number MFCD00132601
    Chemical Name or Material Zeolite ZSM-5, ammonium

    Specifications

    CAS 1318-02-1
    Physical Form Powder
    Surface Area 400m2/g
    Color White
    Quantity 25g
    Avg. Mol. Wt. or Mol. Wt. Range SiO2:Al2O3, 30:1 mole ratio
    MDL Number MFCD00132601
    Chemical Name or Material Zeolite ZSM-5, ammonium

    Description

    • Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit