Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix Shiga Toxin 2-S.cerevisiae Recombinant, Titered
Shop All ZeptoMetrix Corporation Products

Click to view available options
Quantity:
1 mL
Description
- Each frozen aliquot contains 1mL of pure, titered culture of a recombinant strain of Saccharomyces cerevisiae
- Identification of the organism was confirmed by rDNA sequencing
- Biosafety level 1
- Must be used within the confines of Biological Safety cabinet
IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.
Specifications
Specifications
Biosafety Level | 1 |
Certifications/Compliance | ISO, 13485 |
Content And Storage | -65°C |
Form | Solution |
Media Type | Vial |
Product Type | Titered Live Bacteria Control |
For Use With (Application) | Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays |
Quantity | 1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction