Learn More
ZeptoMetrix Immuno-Tek Mouse IgG ELISA Kit

Description
Includes
Microplate, (1x96 well), Detector Antibody (12mL), Mouse IgG Standard (7mL), Assay Diluent (100mL), Plate Wash Buffer (125mL), Substrate (12mL), Stop Solution (12mL), Microtiter Plate Sealers (1pk.), Plastic Bag (1 bag).
Requires
Disposable gloves, Test tubes and racks for preparing specimen and IgG standard dilutions, Validated adjustable micropipettes, single and multi-channel, Distilled or deionized water, Validated incubator capable of maintaining 37° ±1°C, Graduated cylinders and assorted beakers, Validated microtiter plate reader, Automatic microtiter plate washer or manual vacuum aspiration equipment, Timer.
- Contains ready-to-use reagents
- Less performance time
- Microplate and detector antibody in kit specifically balanced to react uniformly with all subclasses of mouse IgG
For monitoring the production and purification of mouse monoclonal antibodies
Specifications
Specifications
Product Type | Immuno-Tek Mouse IgG ELISA kit |
Sufficient For | 96-well Microplate |
For Use With (Application) | Designed for the measurement of Mouse IgG in Plasma, Serum, hybridoma cell supernatants, ascites or other Mouse biological fluids, the kit is especially useful in monito ring the production and purification of Mouse Monoclonal Antibodies |
Kit Contents | 1 plastic bag, 100 mL assay diluent, 12 mL detector antibody, 12 mL stop solution, 12 mL substrate, 125 mL plate wash buffer, 7 mL Human IgG standard, 96-well microplate, Microtiter plate sealers |
Quantity | 96 Determinations |
Storage Requirements | 2°C to 8°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.